Would you like to log in again?
Human Angiostatin K1-3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0545
76237-386EA 723.6 USD InStock
76237-386
Human Angiostatin K1-3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Angiostatin K1-3 AccuSignal ELISA Kit
  • Natural and recombinant human Angiostatin K1-3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: V98-S355(K1-3)
Ang K1-3 is a single, non-glycosylated polypeptide chain containing 259 amino acids. It represents a proteolytic fragment of plasminogen containing the first three kringle structures. Ang K1-3 reduces endothelial cell proliferation and acts as a potent inhibitor of angiogenesis and tumor growth. It displays increased inhibitory activity(ED₅₀ =70 nM) relative to kringles 1-4(ED₅₀ = 135 nM).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Angiostatin K1-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human serum, plasma or cell culture supernates to each empty well. It is recommended that each human Angiostatin K1-3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty