Would you like to log in again?
Human Furin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0553
76237-402EA 635.74 USD InStock
76237-402
Human Furin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Furin AccuSignal ELISA Kit
  • Natural and recombinant human Furin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: D108-E715
Furin(Fur), also known as PACE, is a protein that in humans is encoded by the FURIN gene. The protein encoded by this gene is an enzyme which belongs to the subtilisin-like proprotein convertase family. The FUR gene is located approximately 1 kb upstream of the FES gene on chromosome 15q25-q26. This gene is thought to play a role in tumor progression. Furin is one of the proteases responsible for the proteolytic cleavage of HIV envelope polyprotein precursor gp160 to gp120 and gp41 prior to viral assembly. Furin is enriched in the Golgi apparatus, where it functions to cleave other proteins into their mature/active forms. Expression of furin in T-cells is required for maintenance of peripheral immune tolerance
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human Furin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, tissue homogenates or cell lysates to each empty well. It is recommended that each human Furin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty