Would you like to log in again?
Mouse Angiopoietin-2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0573
76237-434EA 635.74 USD InStock
76237-434
Mouse Angiopoietin-2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Angiopoietin-2 AccuSignal ELISA Kit
  • Natural and recombinant mouse Angiopoietin-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: M1-F496
Angiopoietin-2 is a protein that in humans is encoded by the ANGPTL2 gene. Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. Human and mouse ANGPT2 are 85% identical in amino acid sequence and approximately 60% identical to their ANGPT1 homologs. Transgenic overexpression of ANGPT2 in mice disrupted blood vessel formation in the mouse embryo. By FISH and radiation hybrid analysis, the human ANGPT2 gene was mapped to 8p23, and in a region of syntenic homology on mouse chromosome 8, where the Angpt2 gene was mapped. ANGPT2 is a naturally occurring antagonist of angiopoietin-1 that competes for binding to the TIE2 receptor and blocks ANGPT1-induced TIE2 autophosphorylation during vasculogenesis .
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse Angiopoietin-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse Angiopoietin-2 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty