Would you like to log in again?
Rat Endothelin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0582
76235-704EA 635.74 USD InStock
76235-704
Rat Endothelin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat Endothelin AccuSignal ELISA Kit
  • Natural and recombinant rat Endothelin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: ; Immunogen sequence: C53-W73
The endothelins is a family of structurally and pharmacologically distinct peptides, which has been identified and sequenced in humans. Three isoforms of human endothelin have been identified: endothelin-1, -2, and -3. Endothelin-1 is a potent, 21-amino acid vasoconstrictor peptide produced by vascular endothelial cells. Endothelins are 21-amino acid vasoconstricting peptides produced primarily in the endothelium having a key role in vascular homeostasis. Endothelins are implicated in vascular diseases of several organ systems, including the heart, general circulation and brain. Endothelins are proteins that constrict blood vessels and raise blood pressure. They are normally kept in balance by other mechanisms, but when they are over-expressed, they contribute to high blood pressure(hypertension) and heart disease.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 16.5pg/ml, 7.8pg/ml, 3.9pg/ml rat Endothelin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernatants, serum or plasma(heparin, EDTA) to each empty well. We recommend that each rat Endothelin standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty