Would you like to log in again?
Human Cathepsin S ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0591
76235-720EA 686.86 USD InStock
76235-720
Human Cathepsin S ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Cathepsin S AccuSignal ELISA Kit
  • Natural and recombinant human Cathepsin S.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q17-I331&S109-I331
Cathepsin S, also known as CTSS, is a protein that in humans is encoded by the CTSS gene. A member of the peptidase C1 family, CTSS gene is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. By fluorescence in situ hybridization(FISH), this gene is mapped to 1q21.3. The encoded protein can function as an elastase over a broad pH range in alveolar macrophages. The bond-specificity of this proteinase is in part similar to the specificities of cathepsin L and cathepsin N.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human Cathepsin S standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Cathepsin S standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty