Would you like to log in again?
Human IGFBP6 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0592
76235-722EA 635.74 USD InStock
76235-722
Human IGFBP6 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human IGFBP6 AccuSignal ELISA Kit
  • Natural and recombinant human IGFBP6.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A25-G240
Insulin-like growth factor-binding protein 6(IGFBP6) is a protein that in humans is encoded by the IGFBP6 gene. The IGFBP6 gene is mapped to chromosome 12. In all, it has contained 216 amino acid protein with a calculated molecular mass of 22,847, The human keratinocyte cell line HaCat secretes IGFBP6 as an autocrine growth inhibitor. Recombinant IGFBP6 was also shown to inhibit growth of HaCat cells and other keratinocyte cell lines.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human IGFBP6 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IGFBP6 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty