Would you like to log in again?
Human LYVE-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0597
76235-732EA 687 USD InStock
76235-732
Human LYVE-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human LYVE-1 AccuSignal ELISA Kit
  • Natural and recombinant human LYVE-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S24-T238
Extracellular link domain containing 1, also known as LYVE-1 or XLKD1 is a human gene. The International Radiation Hybrid Mapping Consortium mapped the LYVE1 gene to chromosome 11p15.4. This gene encodes a type I integral membrane glycoprotein. The encoded protein acts as a receptor and binds to both soluble and immobilized hyaluronan. This protein may function in lymphatic hyaluronan transport and have a role in tumor metastasis. This gene may play a role in autocrine regulation of cell growth mediated by growth regulators containing cell surface retention sequence binding(CRS). It may act as a hyaluronan(HA) transporter, either mediating its uptake for catabolism within lymphatic endothelial cells themselves, or its transport into the lumen of afferent lymphatic vessels for subsequent re-uptake and degradation in lymph nodes.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human LYVE-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, cell lysates or tissue homogenates to each empty well. It is recommended that each human LYVE-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty