Would you like to log in again?
Human IL-28A ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0598
76235-734EA 636 USD InStock
76235-734
Human IL-28A ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-28A AccuSignal ELISA Kit
  • Natural and recombinant human IL-28A.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: V26-V200
Interferon lambda-2, also called IFNL2 or IL28A is a class II cytokine receptor ligand related to type I interferons. This gene is mapped to 19q13.2. This gene encodes a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B(IL28B), and interleukin 29(IL29) are three closely related cytokine genes that form a cytokine gene cluster. It up-regulates MHC class I antigen expression. It also displays antitumor activity. The ligand/receptor complex seems to signal through the Jak-STAT pathway. This gene seems not to be essential for early virus-activated host defense in vaginal infection, but plays an important role in Toll-like receptor(TLR)-induced antiviral defense. It seems play a significant role in the antiviral immune defense in the intestinal epithelium.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml human IL-28A standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IL-28A standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty