Would you like to log in again?
Human TNFRSF16 - NGFR ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
KOA0600
76235-738EA 636 USD InStock
76235-738
Human TNFRSF16 - NGFR ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TNFRSF16 - NGFR AccuSignal ELISA Kit
  • Natural and recombinant human TNFRSF16.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: K29-N250
Nerve growth factor receptor (NGFR) is also referred to as p75(NTR) because of its ability to bind at low affinity not only to NGF, but also other neurotrophins including brain-derived neurotrophic factor (BDNF), neurotrophin-3 and neurotrophin-4/5. The nerve growth factor receptor gene is at human chromosome region 17q12-17q22, distal to the chromosome 17 breakpoint in acute leukemias. The neurotrophin receptor p75(NTR), a tumor necrosis factor receptor superfamily member expressed in hepatic stellate cells after fibrotic and cirrhotic liver injury in humans, is a regulator of liver repair. In mice, depletion of p75(NTR) exacerbated liver pathology and inhibited hepatocyte proliferation in vivo. In addition, it is showed that neurotrophins activate p75(NTR) to induce apoptosis through the induction of the sphingomyelin (SM) cycle and increased production of ceramide. Overexpression of p75(NTR) is also found to activate the SM pathway.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human TNFRSF16 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human TNFRSF16 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty