Would you like to log in again?
Human Betacellulin - BTC ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0608
76237-446EA 636 USD InStock
76237-446
Human Betacellulin - BTC ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Betacellulin - BTC AccuSignal ELISA Kit
  • Natural and recombinant human Betacellulin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: D32-Y111
Betacellulin, also known as BTC is a protein that in humans is encoded by the BTC gene. This gene is mapped to 4q13.3. The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor. It is the growth factor that binds to EGFR, ERBB4 and other EGF receptor family members. This protein is a potent mitogen for retinal pigment epithelial cells and vascular smooth muscle cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml human Betacellulin standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human Betacellulin standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty