Would you like to log in again?
Rat FAS-L ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0633
76235-788EA 635.74 USD InStock
76235-788
Rat FAS-L ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat FAS-L AccuSignal ELISA Kit
  • Natural and recombinant rat FAS-L.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q127-L278
FAS Ligand(FASL) is a 40 kDa type II membrane protein belonging to the tumor necrosis factor family, which induces apoptosis by binding to its receptor, Fas. The human FasL gene consists of approximately 8.0 kb and is split into four exons. This gene consists of 281 amino acids with a calculated M(r) of 31,759 and was mapped on chromosome 1q23. It has an identity of 76.9% at the amino acid sequence level with mouse FasL. The FAS and FASL system plays a key role in regulating apoptotic cell death and corruption of this signalling pathway has been shown to participate in immune escape and tumorigenesis. FAS and FASL triggered apoptosis pathway plays an important role in human carcinogenesis. This system may also play a role in modulating the genetic susceptibility of mouse strains to develop T-cell lymphoblastic lymphomas.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat FAS-L standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each rat FAS-L standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty