Would you like to log in again?
Mouse Thrombomodulin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0639
76235-800EA 636 USD InStock
76235-800
Mouse Thrombomodulin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Thrombomodulin AccuSignal ELISA Kit
  • Natural and recombinant mouse Thrombomodulin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L17-S517
Thrombomodulin(TM), CD141 or BDCA-3 is an integral membrane protein expressed on the surface of endothelial cells and serves as a cofactor for thrombin. It reduces blood coagulation by converting thrombin to an anticoagulant enzyme from a procoagulant enzyme. Thrombomodulin is also expressed on human mesothelial cell, monocyte and a dendritic cell subset. In humans, thrombomodulin is encoded by the THBD gene. The protein has a molecular mass of 74kDa, and consists of a single chain with 5 distinct domains. Thrombomodulin functions as a cofactor in the thrombin-induced activation of protein C in the anticoagulant pathway by forming a 1:1 stoichiometric complex with thrombin. This raises the speed of protein C activation thousandfold. Thrombomodulin-bound thrombin has procoagulant effect at the same time by inhibiting fibrinolysis by cleaving thrombin-activatable fibrinolysis inhibitor(TAFI,aka carboxypeptidase B2) into its active form. Thrombomodulin is a glycoprotein on the surface of endothelial cells that, in addition to binding thrombin, regulates C3b inactivation by factor I. Mutations in the thrombomodulin gene(THBD) have also been reported to be associated with atypical hemolytic-uremic syndrome(aHUS).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse Thrombomodulin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin, EDTA, citrate) or urine to each empty well. We recommend that each mouse Thrombomodulin standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty