Would you like to log in again?
Rat IL-17F ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0650
76235-814EA 636 USD InStock
76235-814
Rat IL-17F ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat Interleukin-17F AccuSignal ELISA Kit
  • Natural and recombinant rat IL-17F.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E,coli; Immunogen sequence: A19-A153
Interleukin 17F, also called IL17F is involved in the regulation of normal versus aberrant T-cell responses. This gene is mapped to 6p12.2. The protein encoded by this gene is a cytokine that shares sequence similarity with IL17. This cytokine is expressed by activated T cells, and has been shown to stimulate the production of several other cytokines, including IL6, IL8, and CSF2/GM_CSF. This cytokine is also found to inhibit the angiogenesis of endothelial cells and induce endothelial cells to produce IL2, TGFB1/TGFB, and monocyte chemoattractant protein-1. It is suggested that targeting IL17 and IL17F or antagonizing IL17R might mitigate neutrophil-mediated inflammation in CF.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat IL-17F standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat IL-17F standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty