Would you like to log in again?
Human Granzyme A ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0668
76236-964EA 636 USD InStock
76236-964
Human Granzyme A ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Granzyme A AccuSignal ELISA Kit
  • Natural and recombinant human Granzyme A.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: I29-V262
Granzyme A is a protein that in humans is encoded by the GZMA gene. Cytolytic T lymphocytes (CTL) and natural killer (NK) cells share the remarkable ability to recognize, bind, and lyse specific target cells. They are thought to protect their host by lysing cells bearing on their surface "nonself" antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein described here is a T cell- and natural killer cell-specific serine protease that may function as a common component necessary for lysis of target cells by cytotoxic T lymphocytes and natural killer cells. GZMA induces caspase-independent apoptosis in a characteristic manner, except it causes a distinctive form of DNA damage: single-stranded DNA nicking. A target of GZMA is the SET complex, including HMGB2 and ANP32A.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human Granzyme A standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Granzyme A standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty