Would you like to log in again?
Mouse Periostin - OSF2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0686
76235-852EA 686.86 USD InStock
76235-852
Mouse Periostin - OSF2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Periostin - OSF2 AccuSignal ELISA Kit
  • Natural and recombinant mouse Periostin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: N24-Q836
Periostin, also known as OSF2, is a protein that in humans is encoded by the POSTN gene. The International Radiation Hybrid Mapping Consortium mapped the POSTN gene to chromosome 13. Periostin functions as a ligand for alpha-V/beta-3 and alpha-V/ beta-5 integrins to support adhesion and migration of epithelial cells. It is found that periostin was overexpressed by the majority of human primary breast cancers examined. After myocardial infarction, periostin-induced cardiomyocyte cell cycle reentry and mitosis were associated with improved ventricular remodeling and myocardial function, reduced fibrosis and infarct size, and increase angiogenesis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 6000pg/ml, 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 187.5pg/ml, 93.7pg/m mouse Periostin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse Periostin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty