Would you like to log in again?
Mouse uPAR ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0708
76235-896EA 635.74 USD InStock
76235-896
Mouse uPAR ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse uPAR AccuSignal ELISA Kit
  • Natural and recombinant mouse uPAR.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L24-T297
The urokinase-type plasminogen activator receptor(uPAR) is a key molecule in the regulation of cell-surface plasminogen activation and, as such, plays an important role in many normal as well as pathological processes.1 The cDNA for Mo3, an activation antigen expressed by human monocytes and myelomonocytic cell lines after stimulation by a variety of agents. Mo3 expression in vivo is associated predominantly with macrophages in inflammatory sites. It is a highly glycosylated protein of about 50 kD in monocytes where it is anchored to the plasma membrane by glycosyl-phosphatidylinositol linkage. The complete coding sequence of the cDNA has been found to encode 419 amino acids including a predicted signal peptide of 22 residues and a hydrophobic C-terminal portion. Mo3 is identical to the human receptor for the urokinase plasminogen activator.2 UPAR is a useful prognostic marker for biologically aggressive forms of endometrial cancer.3 PLAUR is located at chromosome 19q13.1-q13.2.1
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse uPAR standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse uPAR standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty