Would you like to log in again?
Human LAP(TGF-beta1) ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0711
76235-902EA 635.74 USD InStock
76235-902
Human LAP(TGF-beta1) ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human LAP(TGF-beta1) AccuSignal ELISA Kit
  • Natural and recombinant human LAP(TGF-beta1).
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L30-R278SF21
TGFbeta1 is secreted as a latent form, which consists of its mature form and a latency-associated peptide(beta1-LAP) in either the presence or the absence of additional latent TGF-beta1-binding protein. Processing and cleavage of the precursor protein between amino acids 278 and 279 results in the formation of LAP dimers and TGF beta dimers that then non-covalently associate with each other to form the small latent TGF beta complex. LAP is secreted and can be found in the extracellular matrix. In addition, LAP can also be expressed on platelets and activated regulatory T cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human LAP(TGF-beta1) standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human LAP(TGF-beta1) standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty