Would you like to log in again?
Mouse CD56 - NCAM1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0716
76235-912EA 686.86 USD InStock
76235-912
Mouse CD56 - NCAM1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CD56 - NCAM1 AccuSignal ELISA Kit
  • Natural and recombinant mouse CD56.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L20-T711
Neural Cell Adhesion Molecule, NCAM, also known as the cluster of differentiation CD56, is a hemophilic binding glycoprotein. It is a glycoprotein of Immunoglobulin(Ig) super family. At least 27 alternatively spliced NCAM mRNAs are produced, giving a wide diversity of NCAM isoforms NCAM gene is located at 11q22-q23.This glycoprotein is mainly expressed on the surface of neurons, glia, skeletal muscle and natural killer cells. NCAM has been implicated as having a role in cell–cell adhesion, neurite outgrowth, synaptic plasticity, and learning and memory.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 40,000pg/ml, 20,000pg/ml, 10,000pg/ml, 5,000pg/ml, 2,500pg/ml, 1,250pg/ml, 625pg/ml mouse CD56 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CD56 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty