Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human TNFRSF14 - HVEM ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0719
76235-918EA 635.74 USD InStock
76235-918
Human TNFRSF14 - HVEM ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TNFRSF14 - HVEM AccuSignal ELISA Kit
  • Natural and recombinant human TNFRSF14.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L39-V202
Tumor necrosis factor receptor superfamily member 14(TNFRSF14), also known as HVEM, is a protein that in humans is encoded by the TNFRSF14 gene. The protein encoded by this gene is a member of the TNF-receptor superfamily. It is mapped to 1p36.32. HVEM plays an important role in HSV pathogenesis because it enhanced the entry of several wildtype HSV strains of both serotypes into CHO cells, and mediated HSV entry into activated human T cells. HVEM and BTLA which are form a bidirectional signaling pathway can regulate cell survival and inhibitory responses between interacting cells. HVEM as an important orchestrator of mucosal immunity integrates signals from innate lymphocytes to induce optimal epithelial Stat3 activation, which indicated that targeting HVEM with agonists could improve host defense.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human TNFRSF14 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human TNFRSF14 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty