Would you like to log in again?
Human SHBG ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0728
76235-936EA 635.74 USD InStock
76235-936
Human SHBG ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human SHBG AccuSignal ELISA Kit
  • Natural and recombinant human SHBG.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L30-H402
Sex hormone-binding globulin (SHBG) or sex steroid-binding globulin (SSBG) is a glycoprotein that binds to the two sex hormones: androgen and estrogen. The gene for SHBG is located on chromosome 17 on the short arm between the bands 17p12→p13. And this gene encodes a steroid binding protein that was first described as a plasma protein secreted by the liver but is now thought to participate in the regulation of steroid responses. The encoded protein transports androgens and estrogens in the blood, binding each steroid molecule as a dimer formed from identical or nearly identical monomers. Polymorphisms in this gene have been associated with polycystic ovary syndrome and type 2 diabetes mellitus. Alternative splicing results in multiple transcript variants.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2nmol/L, 1nmol/L, 0.5nmol/L, 0.25nmol/L, 0.125nmol/L, 0.062nmol/L, 0.031nmol/L human SHBG standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum and plasma(heparin, EDTA) to each empty well. It is recommended that each human SHBG standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty