Would you like to log in again?
Human Renin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0735
76237-466EA 635.74 USD InStock
76237-466
Human Renin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Renin AccuSignal ELISA Kit
  • Natural and recombinant human Renin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L67-R406
Renin, also known as an angiotensinogenase, is an enzyme that participates in the body's renin-angiotensin system(RAS). This gene is mapped to 1q32.1. Renin's primary function is to cause an increase in blood pressure, leading to restoration of perfusion pressure in the kidneys. It also can catalyze the first step in the activation pathway of angiotensinogen--a cascade that can result in aldosterone release,vasoconstriction. Renin, an aspartyl protease, cleaves angiotensinogen to form angiotensin I, which is converted to angiotensin II by angiotensin I converting enzyme, an important regulator of blood pressure and electrolyte balance. What's more, Renin secretion is also stimulated by sympathetic nervous stimulation, mainly through beta-1 adrenoceptor activation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human Renin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each human Renin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty