Would you like to log in again?
Human Thioredoxin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0739
76235-950EA 636 USD InStock
76235-950
Human Thioredoxin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Thioredoxin AccuSignal ELISA Kit
  • Natural and recombinant human Thioredoxin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: V2-V105
Thioredoxin is a class of small redox proteins known to be present in all organisms. It is mapped to 9q31.3. Thioredoxins are proteins that act as antioxidants by facilitating the reduction of other proteins by cysteine thiol-disulfide exchange. They can also act as electron donors to peroxidases and ribonucleotide reductase. Thioredoxin is a 12-kD oxidoreductase enzyme containing a dithiol-disulfide active site. It plays a role in many important biological processes, including redox signaling. This gene also plays a central role in humans and is increasingly linked to medicine through their response to reactive oxygen species (ROS). VDUP1 is a key stress-responsive inhibitor of Thioredoxin activity in cardiomyocytes.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Thioredoxin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human Thioredoxin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty