Would you like to log in again?
Human Ficolin-3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0743
76236-978EA 635.74 USD InStock
76236-978
Human Ficolin-3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Ficolin-3 AccuSignal ELISA Kit
  • Natural and recombinant human Ficolin-3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q24-R299
Ficolin-3, also known as HAKA1, is a protein that in humans is encoded by the FCN3 gene. It is mapped to 1p36.11. The protein can activate the complement pathway in association with MASPs and sMAP, thereby aiding in host defense through the activation of the lectin pathway. Ficolin-3 is expressed as a 35-kD protein that reacts with systemic lupus erythematosus sera. Ficolin-3 does not bind fibronectin(FN1), elastin(ELN), or zymosan, and the lectin activity of the Ficolin-3 is calcium independent. Homozygosity for the variant in the FCN3 gene results in a recessive complement deficiency syndrome.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human Ficolin-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human Ficolin-3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty