Would you like to log in again?
Human DLL1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0752
76235-968EA 636 USD InStock
76235-968
Human DLL1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human DLL1 AccuSignal ELISA Kit
  • Natural and recombinant human DLL1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S22-G540
DLL1(DELTA-LIKE 1), also known as DL1 or DELTA1, is a protein that in humans is encoded by the DLL1 gene. DLL1 is a human homolog of the Notch Delta ligand and is a member of the delta/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis, and it is also important in cell-to-cell communication. This gene is mapped to chromosome 6q27, near a locus for type I diabetes. Functional analysis suggested that a soluble fusion protein containing the DSL domain of DLL1 and its adjacent 50 N-terminal amino acids increased the viability of hemopoietic cells but inhibited cell death. It has been found that DLL1 blocks the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml, 78pg/ml human DLL1 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), urine or human milk to each empty well. We recommend that each human DLL1 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty