Would you like to log in again?
Human IL-18BP ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0758
76235-980EA 649.66 USD InStock
76235-980
Human IL-18BP ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-18BP AccuSignal ELISA Kit
  • Natural and recombinant human IL-18BP.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: T31-G194
Interleukin-18-binding protein, also known as IL18BPa, is a protein that in humans is encoded by the IL18BP gene. It is mapped to 11q13.4. The protein encoded by this gene is an inhibitor of the proinflammatory cytokine IL18. This protein binds to IL18, prevents the binding of IL18 to its receptor, and thus inhibits IL18-induced IFN-gamma production. This protein is constitutively expressed and secreted in mononuclear cells. The expression of this protein can be enhanced by IFN-gamma. It has been found that IL18BP expression in intestinal tissue is increased in endothelial cells as well as cells of the submucosa and overlying lymphoid aggregates in Crohn disease.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 6000pg/ml, 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 187.5pg/ml, 93.7pg/m human IL-18BP standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each human IL-18BP standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty