Would you like to log in again?
Human CRISP3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0760
76235-984EA 635.74 USD InStock
76235-984
Human CRISP3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CRISP3 AccuSignal ELISA Kit
  • Natural and recombinant human CRISP3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: N21-Y245
Cysteine-rich secretory protein 3 is a cysteine-rich secretory protein that in humans is encoded by the CRISP3 gene. CRISP3 expression is found in the secretory epithelium throughout the male genital tract, with particularly high expression in the cauda epididymis and ampulla vas deferens. It is a potential biomarker for prostate cancer. CRISP3 gene can be defined as an early-transcribed gene with a putative role in the pathophysiology of sjogren's syndrome. CRISP3 is present in exocrine secretions and in secretory granules of neutrophilic granulocytes and is believed to play a role in innate immunity.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 100ng/ml, 50ng/ml, 25ng/ml, 1.25ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml human CRISP3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma (heparin, EDTA), saliva, urine or tissue homogenates to each empty well. We recommend that each human CRISP3 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty