Would you like to log in again?
Human CXCL14 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0761
76235-986EA 635.74 USD InStock
76235-986
Human CXCL14 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CXCL14 AccuSignal ELISA Kit
  • Natural and recombinant human CXCL14.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S35-E111
Chemokine(C-X-C motif) ligand 14(CXCL14) is a small cytokine belonging to the CXC chemokine family that is also known as BRAK(for breast and kidney-expressed chemokine). This gene is mapped to 5q31.1. CXCL14 is constitutively expressed at high levels in many normal tissues, it has been found that fibroblasts are the primary source of CXCL14 and that CXCL14 is involved in the homeostasis of monocyte-derived macrophages rather than in inflammation. It is also a potent chemoattractant and activator of dendritic cells, is implicated in homing of these cells, and can stimulate the migration of activated NK cells. CXCL14 also inhibits angiogenesis, possibly as a result of its ability to block endothelial cell chemotaxis. In addition to it, CXCL14 is a potent chemokine for immature DCs, with a chemoattractive activity comparable to bacterial fMLP and MIP1A.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human CXCL14 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CXCL14 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty