Would you like to log in again?
Human CD169 - SIGLEC-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0778
76236-168EA 840 USD InStock
76236-168
Human CD169 - SIGLEC-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD169 - SIGLEC-1 AccuSignal ELISA Kit
  • Natural and recombinant human CD169.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S20-Q1641
SIGLEC-1, also known as Sialoadhesin or CD169, is a cell adhesion molecule found on the surface of certain cells of the immune system called macrophages. This gene is mapped to 20p13. It belongs to the immunoglobulin superfamily(IgSF). Since sialoadhesin binds sialic acids with its N-terminal IgV-domain, it is also a member of the SIGLEC family. The localization and expression of SIGLEC-1 in humans has altered coincident with the evolutionary loss of Neu5Gc. SIGLEC-1-positive macrophages have a dual physiologic function. They act as innate 'flypaper' by preventing the systemic spread of lymph-borne pathogens and as critical gatekeepers at the lymph-tissue interface that facilitate the recognition of particulate antigens by B cells and initiate humoral immune responses.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human CD169 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CD169 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty