Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Mouse TNFSF18 - GITRL ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0782
76236-176EA 635.74 USD InStock
76236-176
Mouse TNFSF18 - GITRL ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse TNFSF18 - GITRL AccuSignal ELISA Kit
  • Natural and recombinant mouse TNFSF18.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: T47-S173
Tumor necrosis factor ligand superfamily member 18, also known as AITRL or GITRL, is a protein that in humans is encoded by the TNFSF18 gene. The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor(TNF) ligand family. It is mapped to 1q25.1. TNFSF18 is a ligand for receptor TNFRSF18/AITR/GITR. It has been shown to modulate T lymphocyte survival in peripheral tissues. This cytokine is also found to be expressed in endothelial cells, and is thought to be important for interaction between T lymphocytes and endothelial cells. TNFSF18-dependent modulation of tryptophan catabolism may represent an important mechanism of action of glucocorticoids, both physiologically and therapeutically.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse TNFSF18 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TNFSF18 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty