Would you like to log in again?
Mouse CD70 - TNFSF7 - cd27L ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0784
76236-180EA 636 USD InStock
76236-180
Mouse CD70 - TNFSF7 - cd27L ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CD70 - TNFSF7 - cd27L AccuSignal ELISA Kit
  • Natural and recombinant mouse CD70.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S45-P195
CD70(Cluster of Differentiation 70), also known as TNFSF7 or CD27L, is a ligand for CD27. This gene is mapped to 19p13.3. CD70 is a surface antigen found on activated, but not resting, T and B lymphocytes. It has been found that CD70 induced proliferation of costimulated T cells and enhanced generation of cytolytic T cells. CD70 is homologous to the ligands of the TNF receptor family, including TNF-alpha, TNF-beta, and the CD40 ligand, showing 19 to 24% amino acid sequence identity in the extracellular region.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse CD70 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CD70 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty