Would you like to log in again?
Human CD21 - CR2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0788
76236-980EA 636 USD InStock
76236-980
Human CD21 - CR2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD21 - CR2 AccuSignal ELISA Kit
  • Natural and recombinant human CD21.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: Ile21-71
Complement receptor type 2(CR2), also known as C3DR or CD21, is a protein that in humans is encoded by the CR2 gene. It is mapped to 1q32.2. CR2 is involved in the complement system. It binds to iC3b(inactive derivative of C3b), C3dg, or C3d. CR2 is the membrane protein on B lymphocytes to which the Epstein-Barr virus (EBV) binds during infection of these cells. CR2, together with CD19, CD81, and CD225, forms the B-cell coreceptor complex, which lowers the activation threshold of the B-cell antigen receptor. Genetic variations in this gene are associated with susceptibility to systemic lupus erythematosus type 9 (SLEB9).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human CD21 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CD21 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty