Would you like to log in again?
Human CD93 - C1qR ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0792
76236-988EA 686.86 USD InStock
76236-988
Human CD93 - C1qR ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD93 - C1qR AccuSignal ELISA Kit
  • Natural and recombinant human CD93.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A24-K580
CD93 (Cluster of Differentiation 93) is a protein that in humans is encoded by the CD93 gene. CD93 belongs to the Group XIV C-Type lectin family and is mapped to 20p11.21. The protein encoded by this gene is a cell-surface glycoprotein and type I membrane protein that was originally identified as a myeloid cell-specific marker. The encoded protein was once thought to be a receptor for C1q, but now is thought to instead be involved in intercellular adhesion and in the clearance of apoptotic cells. The intracellular cytoplasmic tail of this protein has been found to interact with moesin, a protein known to play a role in linking transmembrane proteins to the cytoskeleton and in the remodelling of the cytoskeleton. It has also showed that this receptor is a positive marker for all bone marrow-repopulating stem cells because it is expressed on both CD34-negative and CD34-positive stem cells from umbilical cord blood and adult bone marrow. In addition, highly purified lineage-negative cells with the CD93 protein not only have bone marrow-repopulating capacity but also can differentiate into human hepatocytes in vivo.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml, 78pg/ml human CD93 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma( heparin, EDTA), saliva or urine to each empty well. We recommend that each human CD93 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty