Would you like to log in again?
Human SLAM - CD150 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0793
76236-990EA 635.74 USD InStock
76236-990
Human SLAM - CD150 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human SLAM - CD150 AccuSignal ELISA Kit
  • Natural and recombinant human SLAM.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A21-K236
Signaling lymphocytic activation molecule is a protein that in humans is encoded by the SLAMF1 gene. It belongs to the immunoglobulin gene superfamily. This gene is mapped to 1q23.3. It has found that SLAM is constitutively expressed on peripheral blood memory T cells, T-cell clones, immature thymocytes and a proportion of B cells, and is rapidly induced on naive T cells after activation. In MV-resistant cell lines, infection with clinical MV and expression of SLAM, but not CD46, caused cytopathic effects (CPE). The expression of SLAM on activated B and T lymphocytes correlates with the pathology of MV infection in humans and monkeys, in which lymphoid organs are the chief sites of MV replication and the binding of MV to SLAM may impair the signaling functions of SLAM in lymphocyte activation and inhibit Th0/Th1 cytokine production, thereby promoting Th2 cytokine production. It has reported that antibody-mediated ligation of SLAM on thymocytes triggered a protein tyrosine phosphorylation signal in T cells in a SAP-dependent manner. This signal also involved SHIP, the adaptor molecules DOK2, DOK1, and SHC and RASGAP.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human SLAM standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human SLAM standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty