Would you like to log in again?
Mouse Chemerin - RARRES2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0799
76237-626EA 635.74 USD InStock
76237-626
Mouse Chemerin - RARRES2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Chemerin - RARRES2 AccuSignal ELISA Kit
  • Natural and recombinant mouse Chemerin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: T20-K162
Chemerin, also known as RARRES2 or TIG2, is a protein that is encoded by the RARRES2 gene. It is mapped to 7q36.1. Chemerin is a potent chemoattractant specific for antigen-presenting cells that requires proteolytic activation and acts as a ligand for the G protein-coupled receptor CMKLR1(also known as ChemR23). It is a 14 kDa protein secreted in an inactive form as prochemerin and is activated through cleavage of the C-terminus by inflammatory and coagulation serine proteases. Chemerin was found to stimulate chemotaxis of dendritic cells and macrophages to the site of inflammation. What’s more, the active protein has several roles, including that as an adipokine, and is truncated on both termini from the proprotein.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml mouse Chemerin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse Chemerin standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty