Would you like to log in again?
Human ADAMTS1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0805
76236-198EA 635.74 USD InStock
76236-198
Human ADAMTS1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human ADAMTS1 AccuSignal ELISA Kit
  • Natural and recombinant human ADAMTS1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: F253-A734
ADAMTS1 (A Disintegrin-Like and Metalloproteinase with Thrombospondin Type 1 Motif, 1), also known as METH1, is an enzyme that in humans is encoded by the ADAMTS1 gene. ADAMTS is a family of proteins believed to be anchored to the extracellular matrix (ECM) through interactions with aggregan or other matrix components by one or more thrombospondin type 1 motifs. ADAMTS1 gene is mapped to 21q21.2 based on sequence similarity between the ADAMTS1 sequence and a chromosome 21q21.2 clone. It has been found that ADAMTS1 can disrupt angiogenesis in vivo and in vitro more efficiently than ADAMTS8, THBS1, or endostatin. What’s more, ADAMTS1 disrupts angiogenesis in vivo and in vitro more efficiently than ADAMTS8, THBS1, or endostatin.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 6000pg/ml, 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 187.5pg/ml, 93.7pg/m human ADAMTS1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human ADAMTS1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty