Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Mouse Neuropilin-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0807
76236-202EA 635.74 USD InStock
76236-202
Mouse Neuropilin-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Neuropilin-1 AccuSignal ELISA Kit
  • Natural and recombinant mouse Neuropilin-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: F22-P856
NRP1(Neuropilin 1) also known as NP1, NRP, BDCA4 or VEGF165R, is a membrane-bound coreceptor to a tyrosine kinase receptor for both vascular endothelial growth factor (VEGF) and semaphorin family members. NRP1 plays versatile roles in angiogenesis, axon guidance, cell survival, migration, and invasion. By somatic cell hybrid analysis, the NRP1 gene was mapped to chromosome 10. NRP1 bounds PGF1 with lower affinity. NRP1-mediated interactions are a necessary element in the initiation of the primary immune response and offer another example, like that of agrin, of a molecule shared by neurologic and immunologic synapses. After T-cell contact with DC, T-cell NRP1 colocalized with CD3 in the immunologic synapse and, sometimes, also at the opposite pole of the T cell. Soluble NRP1 interacts in a homophilic fashion with NRP1 on both DC and T cells, and this binding can be inhibited by blocking antibodies to NRP1. Furthermore, selective NRP1 inhibition in this model suppressed neovascular formation substantially.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml mouse Neuropilin-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse Neuropilin-1 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty