Would you like to log in again?
Human Syndecan-4 - SDC4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0819
76237-514EA 636 USD InStock
76237-514
Human Syndecan-4 - SDC4 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Syndecan-4 - SDC4 AccuSignal ELISA Kit
  • Natural and recombinant human Syndecan-4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E19-E145
Syndecan-4 (SDC4) is a protein that in humans is encoded by the SDC4 gene. The protein is found as a homodimer and is a member of the syndecan proteoglycan family. It is mapped to 20q13.12. Syndecan-4 is one of the four vertebrate syndecans and has a molecular weight of ~20 kDa. It is a transmembrane (type I) heparan sulfate proteoglycan that functions as a receptor in intracellular signaling. Syndecan-4 also regulates the actin cytoskeleton, cell adhesion, and cell migration. In addition, Syndecan-4 can activate protein kinase C (PKC). The variable domain of syndecan-4 could be a site of self-association. What’s more, Syndecan-4 also binds to phosphatidylinositol (4,5)-bisphosphate (PIP2) through the variable domain and increases PKC activity ten-fold.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human Syndecan-4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human Syndecan-4 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty