Would you like to log in again?
Human IL-37 - IL1F7 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0824
76236-204EA 636 USD InStock
76236-204
Human IL-37 - IL1F7 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-37 - IL1F7 AccuSignal ELISA Kit
  • Natural and recombinant human IL-37.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: V46-D218
Interleukin 37(IL-37) also named IL1F7, is a protein that in humans is encoded by the IL1F7 gene. The protein encoded by this gene is a member of the interleukin 1 cytokine family. And it is mapped to 2q13. In vitro expression analysis showed that IL-37 was a secretory protein. And IL-37 could play an important role in the link between innate and adaptive immunity and may be useful for tumor immunotherapy. Meanwhile, IL-37 inhibits the production of inflammatory cytokines in peripheral blood mononuclear cells of patients with systemic lupus erythematosus. In addition, IL-37 can emerged as a dual-function cytokine with intra- and extracellular properties for suppressing innate inflammation. Furthermore, IL-37 is a new anti-inflammatory cytokine that plays an important role in protecting against tissue injury during infections via limiting immune and inflammatory reactions.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml human IL-37 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human IL-37 standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty