Would you like to log in again?
Human CD47 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0858
76236-248EA 840 USD InStock
76236-248
Human CD47 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CD47 AccuSignal ELISA Kit
  • Natural and recombinant human CD47.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q19-P139
CD47 (Cluster of Differentiation 47), also known as integrin associated protein (IAP), is a transmembrane protein that in humans is encoded by the CD47 gene. It belongs to the immunoglobulin superfamily and partners with membrane integrins and also binds the ligands thrombospondin-1 (TSP-1) and signal-regulatory protein alpha (SIRPα). CD47 is involved in a range of cellular processes, including apoptosis, proliferation, adhesion, and migration. Furthermore, it plays a key role in immune and angiogenic responses. Also CD47 is ubiquitously expressed in human cells and has found to be overexpressed in many different tumor cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml human CD47 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human CD47 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty