Would you like to log in again?
Ribonuclease T1
Supplier: Rockland Immunochemicals
MB-117-0100
75931-600EA 148.39 USD InStock
75931-600
Ribonuclease T1
Enzymes
Ribonuclease T1
  • Highly purified microbial (non-mammalian) RNase prepared with non-animal components. Supplied as a solution in 2.8M ammonium sulfate.
  • One unit releases the equivalent of one unit A260 of acid-soluble products from yeast RNA in 15 minutes at 37°C at pH 7.5.
Ribonuclease T1 has extensive applications in molecular cloning and DNA sequencing. Because of its specificity it has been a commonly used cleavage enzyme for the determination of structure, nearest neighbor frequencies, and RNA sequencing. The enzyme has further application in the preparation of nucleoside 2',3'-cyclic phosphates, the synthesis of oligonucleotides, and the removal of RNA from DNA preparations. The enzyme is also used as a non-mammalian source of RNase in various applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty