Would you like to log in again?
Diamond DNA Ladder 250 to 25 kbp, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MB-207-0500
RLMB-207-0500EA 143.96 USD InStock
RLMB-207-0500
Diamond DNA Ladder 250 to 25 kbp, Rockland Immunochemicals
Electrophoresis Reagents Molecular Weight Markers
Diamond DNA Ladder 250-25kbp
  • comprised of PCR products and double-stranded DNA
  • digested with restriction enzymes, phenol extracted, equilibrated to 10 mM Tris-HCl (pH 8.0) and 10mM EDTA
Diamond Ready-to-Use DNA Ladder 250-25kbp is a unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 14 fragments, suitable for use as molecular weight standards for agarose gel electrophoresis. The DNA includes fragments ranging from 250-25K base pairs. The 1K and 3K bands have increased intensity to serve as reference points. The approximate mass of DNA in each band is provided (0.52 μg a load) for approximating the mass of DNA in comparably intense samples of similar size.
Diamond Ready-to-Use DNA Ladder 250-25kbp has a suggested load volume is 5 µL/well.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty