Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
12 Epitope MBP Tag Protein Marker Lysate, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MB-301-0100
RLMB-301-0100EA 234.29 USD InStock
RLMB-301-0100
12 Epitope MBP Tag Protein Marker Lysate, Rockland Immunochemicals
Electrophoresis Reagents Molecular Weight Markers
12 Epitope MBP Tag Loading Control Protein Marker
  • Loading control for Western blotting.
  • Lysate expresses 12 different epitope tags.
This product is intended as a positive control for use with anti-epitope tag antibodies.  Epitope tags are short peptide sequences easily recognized by tag-specific antibodies.  Due to their small size, epitope tags do not affect the tagged protein’s biochemical properties.   Most often sequences encoding the epitope tag are included with target DNA at the time of cloning to produce fusion proteins.  This allows anti-epitope tag antibodies to serve as universal detection reagents for any tag containing protein produced by recombinant means.
This product is designed for use as a positive control for anti-epitope tag antibodies. This E.coli lysate contains a recombinant protein comprised of the following commonly used epitope tags: MBP--T7--HSV--cMyc--VSV--Glu-Glu--V5--E-tag--Flag--S tag--HA--6XHis. A 58 kDa recombinant protein is a visualized after SDS-polyacrylamide gel electrophoresis and western blotting using an antibody recognizing any of the expressed epitope tags listed above. The E.coli lysate was prepared by lysing cells in loading buffer/RIPA/Halt protease, then heating to 95° C for 10' followed by centrifugation at 13,000 g for 15'.
Thaw the lysate to room temperature. Gently invert vial several times to ensure that the solution is homogeneous and that any precipitated material is re-dissolved. A loading volume of 0.25 to 2 µL is recommended for western blotting. This product is diluted in 1X SDS-PAGE Sample Buffer (62.5 mM Tris HCL, 2% SDS, 10% Glycerol and 0.005% Bromphenol Blue, pH 6.8).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty