Would you like to log in again?
NPP (1X) ELISA Alkaline Phosphatase Substrate, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
NPP-100
75930-530EA 134 USD InStock
75930-530
NPP (1X) ELISA Alkaline Phosphatase Substrate, Rockland Immunochemicals
Assays ELISAs
(1X) NPP Alkaline Phosphatase ELISA Substrate
  • Buffered in DEA
  • phosphatase based ELISA assays
  • soluble end product
Excellent substrate for alkaline phosphatase based ELISA assays.
Produces a soluble end product which is yellow in color and read at 405nm to 410nm.
Both kinetic and end point determinations can be performed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty