Would you like to log in again?
Rat Peroxidase Anti-Peroxidase (PAP) Soluble Complex, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
P400-0025
75931-480EA 538.82 USD InStock
75931-480
Rat Peroxidase Anti-Peroxidase (PAP) Soluble Complex, Rockland Immunochemicals
Assays Immunological Assays
Rat Peroxidase Anti-Peroxidase (PAP) Soluble Complex
  • peroxidase-anti-peroxidase
  • peroxidase enzymatic labeling
  • amplification of signal, indirectly detecting the amount of primary antibody
The peroxidase-antiperoxidase (PAP) approach utilizes a complex of enzyme and antibody to facilitate amplification of signal, indirectly detecting the amount of primary antibody used in an assay. Generated in Rat, the PAP complex can be utilized in any assay that utilizes a rat primary antibody. With the addition of a secondary antibody that recognizes rat, the reporter PAP complex allows for amplification of the luminescent signal (as three molecules of enzyme are bound to every two immunoglobulins).
Suitable for use in peroxidase enzymatic labeling experiments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty