Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Chicken Blood Defibrinated, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
R102-0050
10800-796EA 323.96 USD InStock
10800-796
Chicken Blood Defibrinated, Rockland Immunochemicals
Blood
Chicken Blood Defibrinated
  • aseptically collected and prepared to order
  • Blood agar plate preparations
Defibrinated chicken blood is aseptically collected and prepared to order from our colony of animals. Fibrin is mechanically removed during the clotting process without the presence of anticoagulants or other additives. Defibrinated chicken blood is ideal for blood agar plate preparations. Blood Agar is a general purpose enriched medium often used to grow fastidious organisms and to differentiate bacteria based on their hemolytic properties. Defibrinated chicken blood is perishable and is collected and processed upon receipt of your order.
Blood agar plate preparations
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty