Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
TURKEY BLOOD in Alsever's, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
R313-0050
RLR313-0050EA 346.53 USD InStock
RLR313-0050
TURKEY BLOOD in Alsever's, Rockland Immunochemicals
Blood
TURKEY BLOOD in Alsevers
  • aseptically collected and prepared to order
  • blood draw in Anticoagulant
Turkey whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Turkey red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty