Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TMB Prestained Red ELISA Peroxidase Substrate, Rockland Immunochemicals

Supplier: Rockland Immunochemicals
TMBE-RED-0100 TMBE-RED-1000
RLTMBE-RED1000EA 318.85 USD InStock
RLTMBE-RED1000 RLTMBE-RED0100
TMB Prestained Red ELISA Peroxidase Substrate, Rockland Immunochemicals
Assays ELISAs
TMB Prestained Red ELISA Peroxidase Substrate

  • pH: 4.0 +/- 0.2
  • Stability @ 18° to 26°C: PASS
  • Stability @ 4°C: PASS
  • QC Raw Material: PASS
  • Absorbance check of final product: PASS
  • Performance Data per ELISA: PASS

TMB ELISA Peroxidase Substrate (3, 3', 5, 5' - Tetramethylbenzidine) is a chromogenic substrate used to visualize antibody reactivity in ELISA experiments. In the presence of peroxidases, TMB can act as an electron donor for the conversion of peroxides to water, typically changing the color of solution to a blue color equivalent to the degree of reactivity and upon addition of acid, changing the solution color to yellow. This proprietary formulation of TMB ELISA Peroxidase Substrate is red in color, enabling users to have visual certainty that the product was applied equally.

TMB ELISA Peroxidase Substrate is ideal for investigators in Immunology, Cancer, and Microbiology research.

TMB Prestained Red ELISA Peroxidase Substrate produces a soluble blue end product of ELISA, read at 370nm or 655nm.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty