Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
TMB Membrane Peroxidase Substrate - Ready-to-Use, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
TMBM-8000 TMBM-20L TMBM-1000
75935-456EA 4031 USD InStock
75935-456 75935-458 75935-454
TMB Membrane Peroxidase Substrate - Ready-to-Use, Rockland Immunochemicals
Assays ELISAs
TMB Membrane Peroxidase Substrate - Ready-To-Use
  • Ready-to-Use
  • insoluble blue-green end product
  • halted with acid to change the TMB to yellow
  • ELISA experiments
TMB Membrane Peroxidase Substrate (3, 3', 5, 5' - Tetramethylbenzidine) is a chromogenic substrate used to visualize antibody reactivity in ELISA experiments. In the presence of peroxidases, TMB can act as an electron donor for the conversion of peroxides to water, changing the color of solution to a blue color equivalent to the degree of reactivity. This reaction can be halted with acid to change the TMB to yellow.
TMB Membrane Peroxidase Substrate is ideal for investigators in Microbiology, Immunology, and Cancer research.
TMB Membrane Peroxidase Substrate will produce an insoluble blue-green end product. Positive signal deposits blue-green color on membrane or surface of sample material. TMB Membrane Peroxidase Substrate incubation time will vary depending on the assay conditions. No dilutions are required, TMB Membrane Peroxidase Substrate comes ready to use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty