Would you like to log in again?
Western Incubation Box, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
WIB-4625-005 WIB-2875-010
75933-630EA 92 USD InStock
75933-630 75933-628
Western Incubation Box, Rockland Immunochemicals
Assays Immunological Assays
Western Incubation Box
  • black plactic incubation box
  • hinged lid closure
  • snap closure
Ideal for Western Blotting using Fluorescent Conjugated Antibodies such as DyLight Conjugated Secondary Antibodies or IRDye Antibody Conjugates.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty