Would you like to log in again?
PBST 10X
Supplier: Rockland Immunochemicals
MB-075-1000
RLMB-075-1000EA 128 USD InStock
RLMB-075-1000
PBST 10X
Buffers Electrophoresis Buffers Washing Buffers
Phosphate Buffer Saline-Tween (PBST) 10X
  • 10X concentrated stock solution
  • 0.2 M Potassium Phosphate, 1.5 M Sodium Chloride and 0.5% (v/v) Tween-20 at a pH of 7.2
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria
  • Use in assay washing procedure
Phosphate Buffer Saline-Tween is commonly use in washing procedure while performing western-blot, ELISA and, more generally immuno-assays.
Application Note: This product is a 10X concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration.  Dilute only prior to immediate use. 10X Phosphate Buffered Saline (PBS) consists of 0.2 M Potassium Phosphate, 1.5 M Sodium Chloride and 0.5% (v/v) Tween-20 at a pH of 7.2. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty